Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140392162-140392435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLGN
Entrez Gene ID
1047
Gene Name
calmegin
Gene Aliases
-
UniGene
Hs.86368
Gene Chromosome Location
Chr.4: 140388453-140427661 on Build GRCh38
Transcripts
NM_001130675.1 NM_004362.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3600450 c.1612G>A p.E538K Chr.4: 140392258-140392258 on Build GRCh38
7234353 c.1523C>A p.T508N Chr.4: 140392347-140392347 on Build GRCh38
3760402 c.1524C>T p.T508T Chr.4: 140392346-140392346 on Build GRCh38
1051443 c.1497A>G p.K499K Chr.4: 140392373-140392373 on Build GRCh38
381541 c.1538C>A p.P513Q Chr.4: 140392332-140392332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34492995 Chr.4: 140392346-140392346 on Build GRCh38
rs755276230 Chr.4: 140392207-140392207 on Build GRCh38
rs763781913 Chr.4: 140392218-140392218 on Build GRCh38
rs753730744 Chr.4: 140392222-140392222 on Build GRCh38
rs746598257 Chr.4: 140392397-140392397 on Build GRCh38
rs543496137 Chr.4: 140392340-140392340 on Build GRCh38
rs774545807 Chr.4: 140392348-140392348 on Build GRCh38
rs767781613 Chr.4: 140392357-140392357 on Build GRCh38
rs750358605 Chr.4: 140392382-140392382 on Build GRCh38
rs773210871 Chr.4: 140392193-140392193 on Build GRCh38
rs145902730 Chr.4: 140392277-140392277 on Build GRCh38
rs75268705 Chr.4: 140392355-140392355 on Build GRCh38
rs997135420 Chr.4: 140392319-140392319 on Build GRCh38
rs370850458 Chr.4: 140392393-140392393 on Build GRCh38
rs910817699 Chr.4: 140392259-140392259 on Build GRCh38
rs376437199 Chr.4: 140392202-140392202 on Build GRCh38
rs754522180 Chr.4: 140392398-140392398 on Build GRCh38
rs943592343 Chr.4: 140392273-140392273 on Build GRCh38
rs745319283 Chr.4: 140392267-140392267 on Build GRCh38
rs879043334 Chr.4: 140392212-140392212 on Build GRCh38
rs768415712 Chr.4: 140392294-140392294 on Build GRCh38
rs765231096 Chr.4: 140392376-140392376 on Build GRCh38
rs116522247 Chr.4: 140392275-140392275 on Build GRCh38
rs549088254 Chr.4: 140392315-140392315 on Build GRCh38
rs757191932 Chr.4: 140392245-140392245 on Build GRCh38
rs772112043 Chr.4: 140392372-140392373 on Build GRCh38
rs779003793 Chr.4: 140392254-140392254 on Build GRCh38
rs758462044 Chr.4: 140392385-140392385 on Build GRCh38
rs776857812 Chr.4: 140392303-140392303 on Build GRCh38
rs893650313 Chr.4: 140392216-140392216 on Build GRCh38
rs984261724 Chr.4: 140392384-140392384 on Build GRCh38
rs370853657 Chr.4: 140392373-140392373 on Build GRCh38
rs774489168 Chr.4: 140392204-140392205 on Build GRCh38
rs770228868 Chr.4: 140392326-140392326 on Build GRCh38
rs753515535 Chr.4: 140392361-140392361 on Build GRCh38
rs751699913 Chr.4: 140392204-140392204 on Build GRCh38
rs1043139889 Chr.4: 140392251-140392251 on Build GRCh38
rs373992784 Chr.4: 140392262-140392263 on Build GRCh38
rs763034849 Chr.4: 140392197-140392197 on Build GRCh38
rs576017421 Chr.4: 140392283-140392283 on Build GRCh38
rs748518042 Chr.4: 140392320-140392320 on Build GRCh38
rs112119780 Chr.4: 140392255-140392255 on Build GRCh38
rs376735755 Chr.4: 140392345-140392345 on Build GRCh38

View Full Product Details