Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109921802-109922049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1
Entrez Gene ID
1435
Gene Name
colony stimulating factor 1
Gene Aliases
CSF-1, MCSF
UniGene
Hs.173894
Gene Chromosome Location
Chr.1: 109910506-109930994 on Build GRCh38
Transcripts
NM_000757.5 NM_172210.2 NM_172211.3 NM_172212.2 XM_017000369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5353462 c.403G>A p.V135I Chr.1: 109921853-109921853 on Build GRCh38
5752898 c.497T>G p.I166S Chr.1: 109921947-109921947 on Build GRCh38
4217619 c.486G>T p.K162N Chr.1: 109921936-109921936 on Build GRCh38
6036820 c.481G>T p.D161Y Chr.1: 109921931-109921931 on Build GRCh38
4217617 c.420G>A p.E140E Chr.1: 109921870-109921870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752598125 Chr.1: 109921825-109921825 on Build GRCh38
rs748611545 Chr.1: 109921882-109921882 on Build GRCh38
rs201377788 Chr.1: 109921931-109921931 on Build GRCh38
rs778137740 Chr.1: 109921985-109921985 on Build GRCh38
rs377085953 Chr.1: 109921853-109921853 on Build GRCh38
rs370213494 Chr.1: 109921857-109921857 on Build GRCh38
rs111936717 Chr.1: 109921883-109921883 on Build GRCh38
rs906002771 Chr.1: 109921827-109921827 on Build GRCh38
rs775583014 Chr.1: 109921920-109921920 on Build GRCh38
rs745478521 Chr.1: 109921859-109921859 on Build GRCh38
rs771489960 Chr.1: 109922002-109922002 on Build GRCh38
rs895842550 Chr.1: 109921944-109921944 on Build GRCh38
rs776825597 Chr.1: 109921911-109921911 on Build GRCh38
rs147128516 Chr.1: 109921990-109921990 on Build GRCh38
rs769778625 Chr.1: 109922010-109922010 on Build GRCh38
rs377701597 Chr.1: 109921858-109921858 on Build GRCh38
rs762780495 Chr.1: 109921924-109921924 on Build GRCh38
rs751503599 Chr.1: 109921932-109921932 on Build GRCh38
rs372834853 Chr.1: 109921829-109921829 on Build GRCh38
rs758709724 Chr.1: 109921984-109921984 on Build GRCh38
rs753443931 Chr.1: 109921982-109921982 on Build GRCh38
rs779131069 Chr.1: 109921879-109921879 on Build GRCh38
rs778192732 Chr.1: 109921895-109921895 on Build GRCh38
rs200625266 Chr.1: 109921971-109921971 on Build GRCh38
rs74387109 Chr.1: 109921973-109921973 on Build GRCh38
rs775733987 Chr.1: 109922011-109922011 on Build GRCh38
rs761881593 Chr.1: 109921936-109921936 on Build GRCh38
rs373767428 Chr.1: 109921840-109921840 on Build GRCh38
rs759668151 Chr.1: 109921915-109921915 on Build GRCh38
rs755839156 Chr.1: 109921877-109921877 on Build GRCh38
rs770123506 Chr.1: 109921916-109921916 on Build GRCh38
rs763299983 Chr.1: 109922024-109922024 on Build GRCh38
rs768541401 Chr.1: 109922025-109922025 on Build GRCh38
rs770954483 Chr.1: 109921903-109921903 on Build GRCh38
rs747532766 Chr.1: 109921902-109921902 on Build GRCh38
rs766913690 Chr.1: 109921948-109921948 on Build GRCh38
rs1057152342 Chr.1: 109921852-109921852 on Build GRCh38
rs575859755 Chr.1: 109921960-109921960 on Build GRCh38
rs764058053 Chr.1: 109921830-109921830 on Build GRCh38
rs781130023 Chr.1: 109922008-109922008 on Build GRCh38

View Full Product Details