Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37496386-37496621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD10
Entrez Gene ID
29775
Gene Name
caspase recruitment domain family member 10
Gene Aliases
BIMP1, CARMA3
UniGene
Hs.57973
Gene Chromosome Location
Chr.22: 37490362-37519203 on Build GRCh38
Transcripts
NM_014550.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7418919 c.1196C>A p.S399* Chr.22: 37496454-37496454 on Build GRCh38
5909868 c.2030C>T p.S677F Chr.22: 37496478-37496478 on Build GRCh38
4890660 c.1953G>T p.R651S Chr.22: 37496555-37496555 on Build GRCh38
3554178 c.2054C>T p.S685L Chr.22: 37496454-37496454 on Build GRCh38
3554177 c.2059+1G>A p.? Chr.22: 37496448-37496448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776672107 Chr.22: 37496563-37496563 on Build GRCh38
rs770909825 Chr.22: 37496559-37496559 on Build GRCh38
rs144994747 Chr.22: 37496522-37496522 on Build GRCh38
rs371707395 Chr.22: 37496428-37496428 on Build GRCh38
rs752877017 Chr.22: 37496574-37496577 on Build GRCh38
rs774751397 Chr.22: 37496438-37496438 on Build GRCh38
rs769110685 Chr.22: 37496528-37496528 on Build GRCh38
rs374827502 Chr.22: 37496412-37496412 on Build GRCh38
rs780289938 Chr.22: 37496424-37496424 on Build GRCh38
rs754598079 Chr.22: 37496406-37496406 on Build GRCh38
rs749725348 Chr.22: 37496516-37496516 on Build GRCh38
rs768665045 Chr.22: 37496483-37496484 on Build GRCh38
rs926427362 Chr.22: 37496576-37496576 on Build GRCh38
rs773438740 Chr.22: 37496546-37496546 on Build GRCh38
rs141109936 Chr.22: 37496475-37496475 on Build GRCh38
rs772296989 Chr.22: 37496450-37496450 on Build GRCh38
rs142113268 Chr.22: 37496454-37496454 on Build GRCh38
rs375211837 Chr.22: 37496429-37496429 on Build GRCh38
rs751008593 Chr.22: 37496499-37496499 on Build GRCh38
rs374348082 Chr.22: 37496416-37496416 on Build GRCh38
rs201152897 Chr.22: 37496543-37496543 on Build GRCh38
rs577751588 Chr.22: 37496529-37496529 on Build GRCh38
rs534915588 Chr.22: 37496440-37496440 on Build GRCh38
rs780670759 Chr.22: 37496515-37496515 on Build GRCh38
rs752343476 Chr.22: 37496413-37496413 on Build GRCh38
rs759371417 Chr.22: 37496582-37496582 on Build GRCh38
rs373196505 Chr.22: 37496481-37496481 on Build GRCh38
rs890360469 Chr.22: 37496585-37496585 on Build GRCh38
rs746396361 Chr.22: 37496417-37496417 on Build GRCh38
rs760692759 Chr.22: 37496461-37496461 on Build GRCh38
rs377261206 Chr.22: 37496484-37496484 on Build GRCh38
rs770299322 Chr.22: 37496422-37496422 on Build GRCh38
rs756596755 Chr.22: 37496504-37496504 on Build GRCh38
rs368935726 Chr.22: 37496414-37496414 on Build GRCh38
rs760865621 Chr.22: 37496555-37496555 on Build GRCh38
rs766281367 Chr.22: 37496464-37496464 on Build GRCh38
rs757997262 Chr.22: 37496492-37496492 on Build GRCh38
rs372414450 Chr.22: 37496480-37496480 on Build GRCh38
rs151150977 Chr.22: 37496469-37496469 on Build GRCh38
rs765039631 Chr.22: 37496598-37496598 on Build GRCh38
rs777296878 Chr.22: 37496497-37496497 on Build GRCh38
rs748408495 Chr.22: 37496536-37496536 on Build GRCh38
rs557461225 Chr.22: 37496602-37496602 on Build GRCh38

View Full Product Details