Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25135884-25136157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf31
Entrez Gene ID
136895
Gene Name
chromosome 7 open reading frame 31
Gene Aliases
-
UniGene
Hs.122055
Gene Chromosome Location
Chr.7: 25134697-25180748 on Build GRCh38
Transcripts
NM_138811.3 XM_005249619.3 XM_005249620.3 XM_011515125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603007 c.1588C>T p.P530S Chr.7: 25136157-25136157 on Build GRCh38
6616571 c.1666C>T p.R556C Chr.7: 25136079-25136079 on Build GRCh38
223149 c.1721C>T p.S574L Chr.7: 25136024-25136024 on Build GRCh38
3637631 c.1755C>T p.S585S Chr.7: 25135990-25135990 on Build GRCh38
5407269 c.1702C>T p.L568F Chr.7: 25136043-25136043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776018553 Chr.7: 25135996-25135996 on Build GRCh38
rs756185943 Chr.7: 25136075-25136075 on Build GRCh38
rs970062712 Chr.7: 25135979-25135979 on Build GRCh38
rs368810357 Chr.7: 25135990-25135990 on Build GRCh38
rs916804112 Chr.7: 25135960-25135960 on Build GRCh38
rs545994696 Chr.7: 25135918-25135919 on Build GRCh38
rs751938580 Chr.7: 25136024-25136024 on Build GRCh38
rs770777340 Chr.7: 25136100-25136100 on Build GRCh38
rs756632034 Chr.7: 25136041-25136041 on Build GRCh38
rs780065462 Chr.7: 25136100-25136101 on Build GRCh38
rs200371370 Chr.7: 25135957-25135957 on Build GRCh38
rs909204147 Chr.7: 25136003-25136003 on Build GRCh38
rs746885791 Chr.7: 25136090-25136090 on Build GRCh38
rs749528242 Chr.7: 25136108-25136110 on Build GRCh38
rs780118788 Chr.7: 25136056-25136056 on Build GRCh38
rs756223414 Chr.7: 25135956-25135956 on Build GRCh38
rs140344221 Chr.7: 25135981-25135981 on Build GRCh38
rs148197323 Chr.7: 25136120-25136120 on Build GRCh38
rs1043022419 Chr.7: 25136135-25136135 on Build GRCh38
rs768769813 Chr.7: 25135967-25135967 on Build GRCh38
rs778768253 Chr.7: 25135973-25135973 on Build GRCh38
rs750886311 Chr.7: 25136037-25136037 on Build GRCh38
rs770518377 Chr.7: 25135995-25135995 on Build GRCh38
rs199628566 Chr.7: 25136131-25136131 on Build GRCh38
rs781705098 Chr.7: 25135949-25135949 on Build GRCh38
rs769464193 Chr.7: 25136122-25136122 on Build GRCh38
rs143176585 Chr.7: 25136079-25136079 on Build GRCh38
rs775042926 Chr.7: 25136126-25136126 on Build GRCh38
rs779055753 Chr.7: 25136078-25136078 on Build GRCh38
rs149543178 Chr.7: 25136032-25136032 on Build GRCh38
rs753936451 Chr.7: 25136064-25136064 on Build GRCh38
rs762213529 Chr.7: 25136029-25136029 on Build GRCh38
rs374917582 Chr.7: 25135952-25135952 on Build GRCh38
rs935072275 Chr.7: 25136013-25136013 on Build GRCh38
rs138018247 Chr.7: 25136023-25136023 on Build GRCh38
rs781296205 Chr.7: 25135955-25135955 on Build GRCh38
rs755217269 Chr.7: 25136075-25136075 on Build GRCh38
rs751874053 Chr.7: 25135927-25135927 on Build GRCh38
rs1036404462 Chr.7: 25136071-25136071 on Build GRCh38
rs535053588 Chr.7: 25136118-25136118 on Build GRCh38
rs749473645 Chr.7: 25135962-25135962 on Build GRCh38
rs757755757 Chr.7: 25135935-25135935 on Build GRCh38
rs1050691411 Chr.7: 25136097-25136097 on Build GRCh38
rs759161975 Chr.7: 25135998-25135998 on Build GRCh38
rs145411485 Chr.7: 25135988-25135988 on Build GRCh38
rs13240851 Chr.7: 25135940-25135940 on Build GRCh38
rs771953633 Chr.7: 25136083-25136083 on Build GRCh38
rs777733476 Chr.7: 25136086-25136086 on Build GRCh38

View Full Product Details