Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140519517-140519790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2A
Entrez Gene ID
27147
Gene Name
DENN domain containing 2A
Gene Aliases
FAM31D, KIAA1277
UniGene
Hs.6385
Gene Chromosome Location
Chr.7: 140518418-140642209 on Build GRCh38
Transcripts
NM_001318052.1 NM_015689.4 NR_134477.1 XM_005249977.3 XM_011516052.2 XM_011516053.2 XM_011516054.2 XM_017011989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471690 c.2916G>A p.L972L Chr.7: 140519714-140519714 on Build GRCh38
4966489 c.2978A>G p.N993S Chr.7: 140519652-140519652 on Build GRCh38
1755123 c.2927G>A p.R976Q Chr.7: 140519703-140519703 on Build GRCh38
6843183 c.2927G>C p.R976P Chr.7: 140519703-140519703 on Build GRCh38
296675 c.2988G>A p.L996L Chr.7: 140519642-140519642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769364711 Chr.7: 140519695-140519695 on Build GRCh38
rs984413767 Chr.7: 140519577-140519577 on Build GRCh38
rs773091139 Chr.7: 140519601-140519601 on Build GRCh38
rs932679862 Chr.7: 140519562-140519562 on Build GRCh38
rs112251962 Chr.7: 140519565-140519565 on Build GRCh38
rs759170730 Chr.7: 140519621-140519621 on Build GRCh38
rs138801925 Chr.7: 140519726-140519726 on Build GRCh38
rs763477958 Chr.7: 140519624-140519624 on Build GRCh38
rs12538386 Chr.7: 140519627-140519627 on Build GRCh38
rs774839406 Chr.7: 140519734-140519734 on Build GRCh38
rs776619198 Chr.7: 140519681-140519681 on Build GRCh38
rs777857876 Chr.7: 140519661-140519661 on Build GRCh38
rs372325499 Chr.7: 140519615-140519615 on Build GRCh38
rs908788988 Chr.7: 140519603-140519603 on Build GRCh38
rs922522144 Chr.7: 140519545-140519545 on Build GRCh38
rs762385988 Chr.7: 140519703-140519703 on Build GRCh38
rs906095259 Chr.7: 140519668-140519668 on Build GRCh38
rs767832417 Chr.7: 140519745-140519745 on Build GRCh38
rs770835290 Chr.7: 140519680-140519680 on Build GRCh38
rs904492767 Chr.7: 140519578-140519578 on Build GRCh38
rs770752162 Chr.7: 140519587-140519588 on Build GRCh38
rs756328923 Chr.7: 140519648-140519648 on Build GRCh38
rs764611339 Chr.7: 140519625-140519625 on Build GRCh38
rs551390715 Chr.7: 140519543-140519543 on Build GRCh38
rs746814060 Chr.7: 140519612-140519612 on Build GRCh38
rs747008356 Chr.7: 140519675-140519675 on Build GRCh38
rs1024601972 Chr.7: 140519665-140519665 on Build GRCh38
rs375723849 Chr.7: 140519660-140519660 on Build GRCh38
rs373925432 Chr.7: 140519638-140519638 on Build GRCh38
rs370003015 Chr.7: 140519690-140519690 on Build GRCh38
rs758420881 Chr.7: 140519761-140519761 on Build GRCh38
rs35301689 Chr.7: 140519674-140519675 on Build GRCh38
rs534591251 Chr.7: 140519622-140519622 on Build GRCh38
rs762369000 Chr.7: 140519743-140519743 on Build GRCh38
rs780137278 Chr.7: 140519652-140519652 on Build GRCh38
rs750619110 Chr.7: 140519639-140519639 on Build GRCh38
rs749345443 Chr.7: 140519659-140519659 on Build GRCh38
rs751868425 Chr.7: 140519626-140519626 on Build GRCh38
rs776241025 Chr.7: 140519597-140519598 on Build GRCh38

View Full Product Details