Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22625341-22625614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK1
Entrez Gene ID
55500
Gene Name
ethanolamine kinase 1
Gene Aliases
EKI, EKI 1, EKI1, Nbla10396
UniGene
Hs.29464
Gene Chromosome Location
Chr.12: 22625142-22690674 on Build GRCh38
Transcripts
NM_001039481.1 NM_018638.4 XM_005253420.4 XM_017019580.1 XM_017019581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5812129 c.232G>A p.V78I Chr.12: 22625395-22625395 on Build GRCh38
5417988 c.179C>T p.T60M Chr.12: 22625342-22625342 on Build GRCh38
4789917 c.394C>T p.H132Y Chr.12: 22625557-22625557 on Build GRCh38
4804142 c.232G>T p.V78F Chr.12: 22625395-22625395 on Build GRCh38
5448422 c.266C>A p.A89D Chr.12: 22625429-22625429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773294981 Chr.12: 22625468-22625468 on Build GRCh38
rs777044008 Chr.12: 22625591-22625591 on Build GRCh38
rs910342753 Chr.12: 22625465-22625465 on Build GRCh38
rs755696439 Chr.12: 22625428-22625429 on Build GRCh38
rs773381371 Chr.12: 22625575-22625575 on Build GRCh38
rs751233420 Chr.12: 22625520-22625520 on Build GRCh38
rs756953756 Chr.12: 22625523-22625523 on Build GRCh38
rs940870303 Chr.12: 22625535-22625535 on Build GRCh38
rs373330052 Chr.12: 22625483-22625483 on Build GRCh38
rs769864803 Chr.12: 22625412-22625412 on Build GRCh38
rs767641778 Chr.12: 22625453-22625453 on Build GRCh38
rs752432601 Chr.12: 22625403-22625404 on Build GRCh38
rs749222898 Chr.12: 22625546-22625546 on Build GRCh38
rs560774822 Chr.12: 22625418-22625418 on Build GRCh38
rs879252894 Chr.12: 22625367-22625367 on Build GRCh38
rs780931224 Chr.12: 22625526-22625526 on Build GRCh38
rs755346854 Chr.12: 22625500-22625500 on Build GRCh38
rs766460988 Chr.12: 22625583-22625583 on Build GRCh38
rs779909625 Chr.12: 22625533-22625533 on Build GRCh38
rs775461746 Chr.12: 22625415-22625415 on Build GRCh38
rs766973112 Chr.12: 22625382-22625383 on Build GRCh38
rs75081525 Chr.12: 22625439-22625439 on Build GRCh38
rs760680188 Chr.12: 22625487-22625487 on Build GRCh38
rs754321450 Chr.12: 22625491-22625491 on Build GRCh38
rs765602646 Chr.12: 22625510-22625510 on Build GRCh38
rs745405516 Chr.12: 22625530-22625530 on Build GRCh38
rs749062164 Chr.12: 22625424-22625424 on Build GRCh38
rs762090798 Chr.12: 22625448-22625448 on Build GRCh38
rs745856468 Chr.12: 22625372-22625372 on Build GRCh38
rs370044157 Chr.12: 22625370-22625370 on Build GRCh38
rs768403929 Chr.12: 22625548-22625548 on Build GRCh38
rs774183018 Chr.12: 22625563-22625563 on Build GRCh38
rs1037336682 Chr.12: 22625476-22625476 on Build GRCh38
rs567759665 Chr.12: 22625455-22625455 on Build GRCh38
rs114417438 Chr.12: 22625574-22625574 on Build GRCh38
rs747881849 Chr.12: 22625568-22625568 on Build GRCh38
rs878870080 Chr.12: 22625389-22625389 on Build GRCh38
rs774480354 Chr.12: 22625445-22625445 on Build GRCh38
rs760764255 Chr.12: 22625579-22625579 on Build GRCh38

View Full Product Details