Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 84229784-84230025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDIL3
Entrez Gene ID
10085
Gene Name
EGF like repeats and discoidin domains 3
Gene Aliases
DEL1
UniGene
Hs.482730
Gene Chromosome Location
Chr.5: 83940595-84384867 on Build GRCh38
Transcripts
NM_001278642.1 NM_005711.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104623 c.226G>T p.G76C Chr.5: 84229855-84229855 on Build GRCh38
1070985 c.212A>G p.E71G Chr.5: 84229869-84229869 on Build GRCh38
3828565 c.208G>A p.E70K Chr.5: 84229873-84229873 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373427567 Chr.5: 84229857-84229857 on Build GRCh38
rs781508019 Chr.5: 84229898-84229898 on Build GRCh38
rs775795701 Chr.5: 84229912-84229912 on Build GRCh38
rs777982656 Chr.5: 84229890-84229890 on Build GRCh38
rs990524053 Chr.5: 84229913-84229913 on Build GRCh38
rs376027513 Chr.5: 84229867-84229867 on Build GRCh38
rs747075601 Chr.5: 84229887-84229889 on Build GRCh38
rs750157489 Chr.5: 84229844-84229844 on Build GRCh38
rs757521094 Chr.5: 84229894-84229894 on Build GRCh38
rs929786026 Chr.5: 84229942-84229942 on Build GRCh38
rs749793907 Chr.5: 84229917-84229917 on Build GRCh38
rs550439681 Chr.5: 84229944-84229944 on Build GRCh38
rs532542202 Chr.5: 84229970-84229970 on Build GRCh38
rs766182006 Chr.5: 84229866-84229866 on Build GRCh38
rs755030558 Chr.5: 84229889-84229889 on Build GRCh38
rs571809768 Chr.5: 84229946-84229946 on Build GRCh38
rs898358931 Chr.5: 84229934-84229934 on Build GRCh38
rs371861139 Chr.5: 84229850-84229850 on Build GRCh38
rs780257935 Chr.5: 84229855-84229857 on Build GRCh38
rs771988127 Chr.5: 84229962-84229962 on Build GRCh38
rs531928677 Chr.5: 84229902-84229902 on Build GRCh38
rs769210309 Chr.5: 84229919-84229919 on Build GRCh38
rs761442329 Chr.5: 84229815-84229815 on Build GRCh38
rs753815487 Chr.5: 84229883-84229883 on Build GRCh38
rs372545376 Chr.5: 84229911-84229911 on Build GRCh38
rs767210264 Chr.5: 84229843-84229843 on Build GRCh38
rs547439718 Chr.5: 84229982-84229982 on Build GRCh38
rs763672716 Chr.5: 84229858-84229858 on Build GRCh38
rs566211766 Chr.5: 84229984-84229984 on Build GRCh38
rs200090780 Chr.5: 84229891-84229891 on Build GRCh38
rs946521169 Chr.5: 84229941-84229941 on Build GRCh38

View Full Product Details