Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21551365-21551590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf51
Entrez Gene ID
339263
Gene Name
chromosome 17 open reading frame 51
Gene Aliases
-
UniGene
Hs.514016
Gene Chromosome Location
Chr.17: 21524606-21551678 on Build GRCh38
Transcripts
NM_001113434.4 XM_005256621.3 XM_006721514.2 XM_011523822.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1588753 c.48C>T p.D16D Chr.17: 21551375-21551375 on Build GRCh38
6427895 c.58G>T p.G20C Chr.17: 21551365-21551365 on Build GRCh38
6846338 c.1A>G p.M1V Chr.17: 21551422-21551422 on Build GRCh38
6866380 c.49G>T p.G17W Chr.17: 21551374-21551374 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182245171 Chr.17: 21551461-21551461 on Build GRCh38
rs563417227 Chr.17: 21551542-21551542 on Build GRCh38
rs782101308 Chr.17: 21551435-21551435 on Build GRCh38
rs545014253 Chr.17: 21551502-21551502 on Build GRCh38
rs577533804 Chr.17: 21551488-21551488 on Build GRCh38
rs782705124 Chr.17: 21551454-21551454 on Build GRCh38
rs541082115 Chr.17: 21551413-21551413 on Build GRCh38
rs567940308 Chr.17: 21551404-21551404 on Build GRCh38
rs374687342 Chr.17: 21551554-21551554 on Build GRCh38

View Full Product Details