Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110327493-110327766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A2
Entrez Gene ID
488
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2
Gene Aliases
ATP2B, DAR, DD, SERCA2
UniGene
Hs.506759
Gene Chromosome Location
Chr.12: 110281227-110351093 on Build GRCh38
Transcripts
NM_001681.3 NM_170665.3 XM_005253888.2 XM_011538402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3416469 c.691G>A p.E231K Chr.12: 110327613-110327613 on Build GRCh38
934938 c.698G>A p.G233D Chr.12: 110327620-110327620 on Build GRCh38
934939 c.767T>G p.F256C Chr.12: 110327689-110327689 on Build GRCh38
3368644 c.690C>T p.T230T Chr.12: 110327612-110327612 on Build GRCh38
4387197 c.811G>T p.V271F Chr.12: 110327733-110327733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767203045 Chr.12: 110327538-110327538 on Build GRCh38
rs986622422 Chr.12: 110327536-110327536 on Build GRCh38
rs757243926 Chr.12: 110327578-110327578 on Build GRCh38
rs549471747 Chr.12: 110327579-110327579 on Build GRCh38
rs778028514 Chr.12: 110327633-110327633 on Build GRCh38
rs143310856 Chr.12: 110327612-110327612 on Build GRCh38
rs531838109 Chr.12: 110327540-110327540 on Build GRCh38
rs771912488 Chr.12: 110327580-110327580 on Build GRCh38
rs748353664 Chr.12: 110327613-110327613 on Build GRCh38
rs778661160 Chr.12: 110327600-110327600 on Build GRCh38
rs376731879 Chr.12: 110327718-110327718 on Build GRCh38
rs369594808 Chr.12: 110327532-110327532 on Build GRCh38
rs944717624 Chr.12: 110327564-110327564 on Build GRCh38
rs142721178 Chr.12: 110327705-110327705 on Build GRCh38
rs750240717 Chr.12: 110327603-110327603 on Build GRCh38
rs759761421 Chr.12: 110327533-110327533 on Build GRCh38
rs188765221 Chr.12: 110327548-110327548 on Build GRCh38
rs1012731102 Chr.12: 110327716-110327716 on Build GRCh38
rs770031210 Chr.12: 110327631-110327631 on Build GRCh38
rs56277015 Chr.12: 110327528-110327528 on Build GRCh38
rs539141956 Chr.12: 110327693-110327693 on Build GRCh38
rs753306264 Chr.12: 110327558-110327558 on Build GRCh38
rs749276909 Chr.12: 110327644-110327644 on Build GRCh38
rs1048975717 Chr.12: 110327666-110327666 on Build GRCh38
rs1043346857 Chr.12: 110327732-110327732 on Build GRCh38
rs760228338 Chr.12: 110327541-110327541 on Build GRCh38
rs121912733 Chr.12: 110327725-110327725 on Build GRCh38

View Full Product Details