Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85100068-85100319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM92B
Entrez Gene ID
339145
Gene Name
family with sequence similarity 92 member B
Gene Aliases
-
UniGene
Hs.125713
Gene Chromosome Location
Chr.16: 85098352-85112508 on Build GRCh38
Transcripts
NM_198491.2 XM_005255931.2 XM_011523063.1 XM_017023198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4809326 c.678T>G p.V226V Chr.16: 85100214-85100214 on Build GRCh38
4941423 c.746C>T p.A249V Chr.16: 85100146-85100146 on Build GRCh38
6016322 c.652-9C>T p.? Chr.16: 85100249-85100249 on Build GRCh38
4914021 c.698G>A p.R233Q Chr.16: 85100194-85100194 on Build GRCh38
5385508 c.725C>T p.P242L Chr.16: 85100167-85100167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748774998 Chr.16: 85100127-85100127 on Build GRCh38
rs962954913 Chr.16: 85100118-85100118 on Build GRCh38
rs371033693 Chr.16: 85100272-85100272 on Build GRCh38
rs142788358 Chr.16: 85100208-85100208 on Build GRCh38
rs959181717 Chr.16: 85100151-85100151 on Build GRCh38
rs1053444007 Chr.16: 85100247-85100247 on Build GRCh38
rs377580020 Chr.16: 85100095-85100095 on Build GRCh38
rs761814834 Chr.16: 85100278-85100278 on Build GRCh38
rs769829363 Chr.16: 85100117-85100117 on Build GRCh38
rs891449399 Chr.16: 85100097-85100097 on Build GRCh38
rs1054949189 Chr.16: 85100089-85100089 on Build GRCh38
rs768415956 Chr.16: 85100190-85100190 on Build GRCh38
rs747703011 Chr.16: 85100199-85100199 on Build GRCh38
rs1026316867 Chr.16: 85100297-85100297 on Build GRCh38
rs779569445 Chr.16: 85100175-85100175 on Build GRCh38
rs147408036 Chr.16: 85100211-85100211 on Build GRCh38
rs751596851 Chr.16: 85100235-85100235 on Build GRCh38
rs746347583 Chr.16: 85100267-85100267 on Build GRCh38
rs192144400 Chr.16: 85100273-85100273 on Build GRCh38
rs759998712 Chr.16: 85100147-85100147 on Build GRCh38
rs779033165 Chr.16: 85100098-85100098 on Build GRCh38
rs780941023 Chr.16: 85100116-85100116 on Build GRCh38
rs533752878 Chr.16: 85100136-85100136 on Build GRCh38
rs201114826 Chr.16: 85100173-85100173 on Build GRCh38
rs368494980 Chr.16: 85100288-85100288 on Build GRCh38
rs376462802 Chr.16: 85100092-85100092 on Build GRCh38
rs201630543 Chr.16: 85100132-85100132 on Build GRCh38
rs557924096 Chr.16: 85100126-85100126 on Build GRCh38
rs3764278 Chr.16: 85100266-85100266 on Build GRCh38
rs752863344 Chr.16: 85100148-85100148 on Build GRCh38
rs553906448 Chr.16: 85100156-85100156 on Build GRCh38
rs116395098 Chr.16: 85100224-85100224 on Build GRCh38
rs759875228 Chr.16: 85100213-85100213 on Build GRCh38
rs771710204 Chr.16: 85100264-85100264 on Build GRCh38
rs981670277 Chr.16: 85100195-85100195 on Build GRCh38
rs778742106 Chr.16: 85100258-85100258 on Build GRCh38
rs747601319 Chr.16: 85100261-85100261 on Build GRCh38
rs751686547 Chr.16: 85100170-85100170 on Build GRCh38
rs773978785 Chr.16: 85100191-85100191 on Build GRCh38
rs918628592 Chr.16: 85100154-85100154 on Build GRCh38
rs779705252 Chr.16: 85100246-85100246 on Build GRCh38
rs560771176 Chr.16: 85100286-85100286 on Build GRCh38
rs969745301 Chr.16: 85100110-85100110 on Build GRCh38
rs202130672 Chr.16: 85100289-85100289 on Build GRCh38
rs374156071 Chr.16: 85100102-85100102 on Build GRCh38
rs145807953 Chr.16: 85100111-85100111 on Build GRCh38
rs781141108 Chr.16: 85100171-85100171 on Build GRCh38
rs1017084627 Chr.16: 85100277-85100277 on Build GRCh38
rs775981140 Chr.16: 85100226-85100226 on Build GRCh38
rs373006193 Chr.16: 85100133-85100133 on Build GRCh38
rs764416082 Chr.16: 85100231-85100231 on Build GRCh38
rs148969047 Chr.16: 85100179-85100179 on Build GRCh38
rs767045254 Chr.16: 85100138-85100138 on Build GRCh38
rs745367591 Chr.16: 85100112-85100112 on Build GRCh38
rs373626017 Chr.16: 85100249-85100249 on Build GRCh38
rs757352501 Chr.16: 85100236-85100236 on Build GRCh38
rs777107768 Chr.16: 85100139-85100139 on Build GRCh38
rs370506826 Chr.16: 85100099-85100099 on Build GRCh38
rs767386202 Chr.16: 85100243-85100243 on Build GRCh38
rs374703386 Chr.16: 85100244-85100244 on Build GRCh38
rs544191027 Chr.16: 85100254-85100254 on Build GRCh38
rs764066698 Chr.16: 85100166-85100166 on Build GRCh38
rs149810857 Chr.16: 85100093-85100093 on Build GRCh38
rs905638293 Chr.16: 85100270-85100270 on Build GRCh38
rs546892318 Chr.16: 85100276-85100276 on Build GRCh38
rs1017033758 Chr.16: 85100113-85100113 on Build GRCh38
rs368759847 Chr.16: 85100245-85100245 on Build GRCh38
rs748892560 Chr.16: 85100184-85100184 on Build GRCh38
rs763063064 Chr.16: 85100230-85100230 on Build GRCh38

View Full Product Details