Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200605473-200605698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX1
Entrez Gene ID
316
Gene Name
aldehyde oxidase 1
Gene Aliases
AO, AOH1
UniGene
Hs.406238
Gene Chromosome Location
Chr.2: 200585952-200677064 on Build GRCh38
Transcripts
NM_001159.3 XM_011511062.1 XM_017003946.1 XM_017003947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183092 c.869G>A p.R290K Chr.2: 200605590-200605590 on Build GRCh38
4764357 c.883A>C p.S295R Chr.2: 200605604-200605604 on Build GRCh38
7336459 c.865G>A p.D289N Chr.2: 200605586-200605586 on Build GRCh38
4944524 c.828A>G p.K276K Chr.2: 200605549-200605549 on Build GRCh38
6399017 c.844C>T p.H282Y Chr.2: 200605565-200605565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373426863 Chr.2: 200605559-200605559 on Build GRCh38
rs550377398 Chr.2: 200605512-200605515 on Build GRCh38
rs760576452 Chr.2: 200605564-200605564 on Build GRCh38
rs759457645 Chr.2: 200605523-200605526 on Build GRCh38
rs201023748 Chr.2: 200605523-200605523 on Build GRCh38
rs773865710 Chr.2: 200605557-200605557 on Build GRCh38
rs869035962 Chr.2: 200605519-200605522 on Build GRCh38
rs772716079 Chr.2: 200605561-200605561 on Build GRCh38
rs200355852 Chr.2: 200605537-200605537 on Build GRCh38
rs558974283 Chr.2: 200605514-200605515 on Build GRCh38
rs760348428 Chr.2: 200605507-200605507 on Build GRCh38
rs781753651 Chr.2: 200605551-200605551 on Build GRCh38
rs554846029 Chr.2: 200605513-200605515 on Build GRCh38
rs374138035 Chr.2: 200605666-200605666 on Build GRCh38
rs754142864 Chr.2: 200605512-200605514 on Build GRCh38
rs764126716 Chr.2: 200605524-200605526 on Build GRCh38
rs758712449 Chr.2: 200605529-200605529 on Build GRCh38
rs869208204 Chr.2: 200605523-200605525 on Build GRCh38
rs752763068 Chr.2: 200605528-200605528 on Build GRCh38
rs371230713 Chr.2: 200605665-200605665 on Build GRCh38
rs201739752 Chr.2: 200605633-200605633 on Build GRCh38
rs560343253 Chr.2: 200605515-200605516 on Build GRCh38
rs199651858 Chr.2: 200605511-200605512 on Build GRCh38
rs879107907 Chr.2: 200605513-200605514 on Build GRCh38
rs753966652 Chr.2: 200605526-200605527 on Build GRCh38
rs866607614 Chr.2: 200605508-200605508 on Build GRCh38
rs776419502 Chr.2: 200605583-200605583 on Build GRCh38
rs753703328 Chr.2: 200605520-200605520 on Build GRCh38
rs945339648 Chr.2: 200605558-200605558 on Build GRCh38
rs766081863 Chr.2: 200605569-200605569 on Build GRCh38
rs377374339 Chr.2: 200605567-200605567 on Build GRCh38
rs61280547 Chr.2: 200605573-200605573 on Build GRCh38
rs3038607 Chr.2: 200605511-200605511 on Build GRCh38
rs200778985 Chr.2: 200605511-200605513 on Build GRCh38
rs546168308 Chr.2: 200605511-200605515 on Build GRCh38
rs752850873 Chr.2: 200605628-200605628 on Build GRCh38
rs562795689 Chr.2: 200605515-200605515 on Build GRCh38
rs770220813 Chr.2: 200605556-200605556 on Build GRCh38
rs1004353665 Chr.2: 200605588-200605588 on Build GRCh38
rs796289738 Chr.2: 200605522-200605525 on Build GRCh38
rs368170603 Chr.2: 200605674-200605674 on Build GRCh38
rs879086669 Chr.2: 200605514-200605514 on Build GRCh38
rs766163757 Chr.2: 200605510-200605511 on Build GRCh38
rs759556652 Chr.2: 200605609-200605609 on Build GRCh38
rs755173670 Chr.2: 200605525-200605525 on Build GRCh38
rs779136370 Chr.2: 200605526-200605526 on Build GRCh38
rs371355321 Chr.2: 200605622-200605622 on Build GRCh38
rs757710099 Chr.2: 200605634-200605634 on Build GRCh38
rs757418195 Chr.2: 200605527-200605528 on Build GRCh38
rs746371880 Chr.2: 200605553-200605553 on Build GRCh38
rs200252546 Chr.2: 200605522-200605522 on Build GRCh38
rs57205763 Chr.2: 200605511-200605514 on Build GRCh38
rs73049078 Chr.2: 200605631-200605631 on Build GRCh38
rs747444891 Chr.2: 200605536-200605536 on Build GRCh38

View Full Product Details