Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22078352-22078625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42
Entrez Gene ID
998
Gene Name
cell division cycle 42
Gene Aliases
CDC42Hs, G25K, TKS
UniGene
Hs.467637
Gene Chromosome Location
Chr.1: 22052627-22092943 on Build GRCh38
Transcripts
NM_001039802.1 NM_001791.3 NM_044472.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
46468 c.89C>T p.S30L Chr.1: 22078567-22078567 on Build GRCh38
1247757 c.30C>T p.G10G Chr.1: 22078508-22078508 on Build GRCh38
6256110 c.44G>T p.G15V Chr.1: 22078522-22078522 on Build GRCh38
3733072 c.39C>T p.I13I Chr.1: 22078567-22078567 on Build GRCh38
6337850 c.30C>T p.G10G Chr.1: 22078508-22078508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965447224 Chr.1: 22078536-22078536 on Build GRCh38
rs745673041 Chr.1: 22078505-22078505 on Build GRCh38
rs776809649 Chr.1: 22078580-22078580 on Build GRCh38
rs777290250 Chr.1: 22078453-22078453 on Build GRCh38
rs575366184 Chr.1: 22078405-22078405 on Build GRCh38
rs542688819 Chr.1: 22078431-22078431 on Build GRCh38
rs373664682 Chr.1: 22078458-22078458 on Build GRCh38
rs16826470 Chr.1: 22078433-22078433 on Build GRCh38
rs765830067 Chr.1: 22078590-22078590 on Build GRCh38
rs375924804 Chr.1: 22078508-22078508 on Build GRCh38
rs775718828 Chr.1: 22078529-22078529 on Build GRCh38
rs750902467 Chr.1: 22078595-22078595 on Build GRCh38
rs1019874387 Chr.1: 22078489-22078489 on Build GRCh38
rs769221393 Chr.1: 22078442-22078442 on Build GRCh38
rs761795033 Chr.1: 22078583-22078583 on Build GRCh38
rs572342695 Chr.1: 22078419-22078419 on Build GRCh38
rs902035547 Chr.1: 22078400-22078400 on Build GRCh38
rs1026869158 Chr.1: 22078426-22078426 on Build GRCh38
rs760045099 Chr.1: 22078470-22078470 on Build GRCh38
rs770945327 Chr.1: 22078463-22078463 on Build GRCh38
rs797044916 Chr.1: 22078546-22078546 on Build GRCh38
rs76851854 Chr.1: 22078386-22078386 on Build GRCh38
rs760835408 Chr.1: 22078565-22078565 on Build GRCh38
rs761486028 Chr.1: 22078583-22078584 on Build GRCh38
rs774271691 Chr.1: 22078484-22078484 on Build GRCh38
rs368993116 Chr.1: 22078568-22078568 on Build GRCh38

View Full Product Details