Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 229366676-229366935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNER
Entrez Gene ID
92737
Gene Name
delta/notch like EGF repeat containing
Gene Aliases
UNQ26, bet
UniGene
Hs.234074
Gene Chromosome Location
Chr.2: 229357629-229714570 on Build GRCh38
Transcripts
NM_139072.3 XM_005246950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4168815 c.2092C>T p.R698W Chr.2: 229366883-229366883 on Build GRCh38
7250397 c.2093G>T p.R698L Chr.2: 229366882-229366882 on Build GRCh38
3364626 c.2050C>A p.R684S Chr.2: 229366925-229366925 on Build GRCh38
4913880 c.2064C>A p.S688R Chr.2: 229366911-229366911 on Build GRCh38
1017501 c.2083G>A p.A695T Chr.2: 229366892-229366892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044249087 Chr.2: 229366723-229366723 on Build GRCh38
rs751453763 Chr.2: 229366846-229366846 on Build GRCh38
rs139514662 Chr.2: 229366759-229366759 on Build GRCh38
rs775072382 Chr.2: 229366884-229366884 on Build GRCh38
rs774872655 Chr.2: 229366720-229366720 on Build GRCh38
rs758506865 Chr.2: 229366859-229366859 on Build GRCh38
rs1019207998 Chr.2: 229366789-229366789 on Build GRCh38
rs150035332 Chr.2: 229366910-229366910 on Build GRCh38
rs748951182 Chr.2: 229366892-229366892 on Build GRCh38
rs778014620 Chr.2: 229366864-229366864 on Build GRCh38
rs772826749 Chr.2: 229366857-229366857 on Build GRCh38
rs762406084 Chr.2: 229366842-229366842 on Build GRCh38
rs948558995 Chr.2: 229366708-229366708 on Build GRCh38
rs767334705 Chr.2: 229366900-229366900 on Build GRCh38
rs367716187 Chr.2: 229366908-229366908 on Build GRCh38
rs112975208 Chr.2: 229366854-229366854 on Build GRCh38
rs761443379 Chr.2: 229366897-229366897 on Build GRCh38
rs763615331 Chr.2: 229366844-229366844 on Build GRCh38
rs752912056 Chr.2: 229366917-229366917 on Build GRCh38
rs769410792 Chr.2: 229366828-229366828 on Build GRCh38
rs770496301 Chr.2: 229366752-229366752 on Build GRCh38
rs898996837 Chr.2: 229366733-229366733 on Build GRCh38
rs770173487 Chr.2: 229366883-229366883 on Build GRCh38
rs576440212 Chr.2: 229366704-229366704 on Build GRCh38
rs145368755 Chr.2: 229366916-229366916 on Build GRCh38
rs557868811 Chr.2: 229366703-229366703 on Build GRCh38
rs768087498 Chr.2: 229366894-229366894 on Build GRCh38
rs750171761 Chr.2: 229366855-229366855 on Build GRCh38
rs747487250 Chr.2: 229366865-229366865 on Build GRCh38
rs775160268 Chr.2: 229366830-229366830 on Build GRCh38
rs760016453 Chr.2: 229366825-229366825 on Build GRCh38
rs771341328 Chr.2: 229366867-229366867 on Build GRCh38
rs111685443 Chr.2: 229366881-229366881 on Build GRCh38
rs373612462 Chr.2: 229366858-229366858 on Build GRCh38
rs761660347 Chr.2: 229366853-229366853 on Build GRCh38
rs143853534 Chr.2: 229366882-229366882 on Build GRCh38
rs60699589 Chr.2: 229366756-229366756 on Build GRCh38
rs778522656 Chr.2: 229366895-229366895 on Build GRCh38
rs200724319 Chr.2: 229366915-229366915 on Build GRCh38
rs111775882 Chr.2: 229366790-229366790 on Build GRCh38
rs74488906 Chr.2: 229366791-229366791 on Build GRCh38
rs762238880 Chr.2: 229366736-229366736 on Build GRCh38

View Full Product Details