Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25427416-25427683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
UniGene
Hs.307720
Gene Chromosome Location
Chr.2: 25377220-25673647 on Build GRCh38
Transcripts
NM_001256303.1 NM_001256304.1 NM_001256308.2 NM_001320932.1 NM_001320933.1 NM_001320934.1 NM_001320935.1 NM_001320936.1 NM_021907.4 NM_033147.3 NM_033148.3 NM_183360.2 NM_183361.2 XM_005264182.1 XM_011532671.1 XM_011532672.2 XM_011532673.1 XM_011532674.1 XM_011532675.1 XM_011532676.1 XM_011532677.2 XM_011532679.1 XM_011532680.2 XM_011532681.2 XM_017003529.1 XM_017003530.1 XM_017003531.1 XM_017003532.1 XM_017003533.1 XM_017003534.1 XM_017003535.1 XM_017003536.1 XM_017003537.1 XM_017003538.1 XM_017003539.1 XM_017003540.1 XM_017003541.1 XM_017003542.1 XM_017003543.1 XM_017003544.1 XM_017003545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650031 c.1510C>T p.R504W Chr.2: 25427579-25427579 on Build GRCh38
4093277 c.1490C>T p.S497L Chr.2: 25427599-25427599 on Build GRCh38
4093278 c.1490C>T p.S497L Chr.2: 25427599-25427599 on Build GRCh38
6650030 c.1510C>T p.R504W Chr.2: 25427579-25427579 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs189786450 Chr.2: 25427485-25427485 on Build GRCh38
rs768077118 Chr.2: 25427550-25427550 on Build GRCh38
rs368115598 Chr.2: 25427599-25427599 on Build GRCh38
rs768015290 Chr.2: 25427577-25427577 on Build GRCh38
rs754314448 Chr.2: 25427584-25427584 on Build GRCh38
rs755000320 Chr.2: 25427612-25427612 on Build GRCh38
rs943729937 Chr.2: 25427491-25427491 on Build GRCh38
rs557301120 Chr.2: 25427474-25427474 on Build GRCh38
rs374448659 Chr.2: 25427516-25427516 on Build GRCh38
rs775709324 Chr.2: 25427643-25427643 on Build GRCh38
rs929271656 Chr.2: 25427589-25427589 on Build GRCh38
rs779889030 Chr.2: 25427490-25427490 on Build GRCh38
rs746769895 Chr.2: 25427497-25427497 on Build GRCh38
rs567750169 Chr.2: 25427463-25427463 on Build GRCh38
rs370287747 Chr.2: 25427524-25427524 on Build GRCh38
rs780906020 Chr.2: 25427615-25427615 on Build GRCh38
rs756914598 Chr.2: 25427586-25427586 on Build GRCh38
rs768304008 Chr.2: 25427515-25427515 on Build GRCh38
rs557137537 Chr.2: 25427578-25427578 on Build GRCh38
rs745665110 Chr.2: 25427492-25427494 on Build GRCh38
rs373630664 Chr.2: 25427576-25427576 on Build GRCh38
rs764987873 Chr.2: 25427595-25427595 on Build GRCh38
rs772018018 Chr.2: 25427642-25427642 on Build GRCh38
rs749913189 Chr.2: 25427596-25427596 on Build GRCh38
rs746146564 Chr.2: 25427638-25427638 on Build GRCh38
rs746915944 Chr.2: 25427609-25427609 on Build GRCh38
rs1000549675 Chr.2: 25427442-25427442 on Build GRCh38
rs943299905 Chr.2: 25427633-25427633 on Build GRCh38
rs199722054 Chr.2: 25427598-25427598 on Build GRCh38
rs539774498 Chr.2: 25427650-25427650 on Build GRCh38
rs771099430 Chr.2: 25427623-25427623 on Build GRCh38
rs774575754 Chr.2: 25427628-25427628 on Build GRCh38
rs771152834 Chr.2: 25427525-25427525 on Build GRCh38
rs75591229 Chr.2: 25427460-25427460 on Build GRCh38
rs774345423 Chr.2: 25427530-25427530 on Build GRCh38
rs761287975 Chr.2: 25427579-25427579 on Build GRCh38
rs747939462 Chr.2: 25427621-25427621 on Build GRCh38
rs947172062 Chr.2: 25427443-25427443 on Build GRCh38
rs771814720 Chr.2: 25427608-25427609 on Build GRCh38

View Full Product Details