Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46618764-46619023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIPT
Entrez Gene ID
9419
Gene Name
CXXC repeat containing interactor of PDZ3 domain
Gene Aliases
HSPC139, SSMDF
UniGene
Hs.133998
Gene Chromosome Location
Chr.2: 46617172-46630176 on Build GRCh38
Transcripts
NM_014171.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668895 c.40A>T p.I14F Chr.2: 46618796-46618796 on Build GRCh38
7314707 c.72_73insA p.N25fs*12 Chr.2: 46618828-46618829 on Build GRCh38
6635516 c.33T>C p.G11G Chr.2: 46618789-46618789 on Build GRCh38
6579672 c.17-4A>T p.? Chr.2: 46618769-46618769 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs570169344 Chr.2: 46618927-46618927 on Build GRCh38
rs145115368 Chr.2: 46618825-46618825 on Build GRCh38
rs748425635 Chr.2: 46618852-46618852 on Build GRCh38
rs747435551 Chr.2: 46618860-46618860 on Build GRCh38
rs750253997 Chr.2: 46618806-46618806 on Build GRCh38
rs778110670 Chr.2: 46618859-46618859 on Build GRCh38
rs755092497 Chr.2: 46618833-46618833 on Build GRCh38
rs1050581025 Chr.2: 46618949-46618949 on Build GRCh38
rs758511452 Chr.2: 46618854-46618854 on Build GRCh38
rs768231925 Chr.2: 46618880-46618880 on Build GRCh38
rs555129716 Chr.2: 46618918-46618918 on Build GRCh38
rs142774632 Chr.2: 46618867-46618867 on Build GRCh38
rs534089986 Chr.2: 46618905-46618905 on Build GRCh38
rs771417832 Chr.2: 46618861-46618861 on Build GRCh38
rs1009941521 Chr.2: 46618814-46618814 on Build GRCh38
rs1046370125 Chr.2: 46618893-46618893 on Build GRCh38
rs755877821 Chr.2: 46618808-46618808 on Build GRCh38
rs746606165 Chr.2: 46618981-46618981 on Build GRCh38
rs117720191 Chr.2: 46618992-46618992 on Build GRCh38
rs753774289 Chr.2: 46618827-46618827 on Build GRCh38
rs778804243 Chr.2: 46618841-46618841 on Build GRCh38
rs373483934 Chr.2: 46618834-46618834 on Build GRCh38
rs368976360 Chr.2: 46618872-46618872 on Build GRCh38
rs146957186 Chr.2: 46618894-46618894 on Build GRCh38
rs571356589 Chr.2: 46618897-46618897 on Build GRCh38
rs149286673 Chr.2: 46618798-46618798 on Build GRCh38
rs1009580431 Chr.2: 46618984-46618984 on Build GRCh38
rs917778288 Chr.2: 46618988-46618988 on Build GRCh38
rs950022903 Chr.2: 46618887-46618887 on Build GRCh38
rs769470745 Chr.2: 46618829-46618829 on Build GRCh38
rs899296740 Chr.2: 46618934-46618934 on Build GRCh38
rs767328883 Chr.2: 46618797-46618797 on Build GRCh38
rs367574133 Chr.2: 46618990-46618990 on Build GRCh38
rs926341839 Chr.2: 46618971-46618971 on Build GRCh38
rs937763505 Chr.2: 46618982-46618983 on Build GRCh38
rs777301811 Chr.2: 46618941-46618941 on Build GRCh38

View Full Product Details