Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 10212438-10212711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN4
Entrez Gene ID
1183
Gene Name
chloride voltage-gated channel 4
Gene Aliases
CLC4, ClC-4, ClC-4A
UniGene
Hs.495674
Gene Chromosome Location
Chr.X: 10156945-10237660 on Build GRCh38
Transcripts
NM_001256944.1 NM_001830.3

SNP Detail

SNP ID Chromosome Location
rs200303958 Chr.X: 10212596-10212596 on Build GRCh38
rs903970026 Chr.X: 10212663-10212663 on Build GRCh38
rs756555007 Chr.X: 10212680-10212680 on Build GRCh38
rs112652769 Chr.X: 10212652-10212652 on Build GRCh38
rs150240935 Chr.X: 10212472-10212472 on Build GRCh38
rs376621307 Chr.X: 10212478-10212478 on Build GRCh38
rs141271427 Chr.X: 10212463-10212463 on Build GRCh38
rs372538958 Chr.X: 10212687-10212687 on Build GRCh38
rs751550003 Chr.X: 10212671-10212671 on Build GRCh38
rs948007734 Chr.X: 10212657-10212657 on Build GRCh38
rs34148950 Chr.X: 10212514-10212514 on Build GRCh38
rs775103186 Chr.X: 10212643-10212643 on Build GRCh38
rs766499844 Chr.X: 10212668-10212668 on Build GRCh38
rs918406808 Chr.X: 10212604-10212604 on Build GRCh38
rs767324550 Chr.X: 10212490-10212490 on Build GRCh38
rs775225884 Chr.X: 10212505-10212505 on Build GRCh38
rs757664981 Chr.X: 10212686-10212686 on Build GRCh38
rs781711653 Chr.X: 10212539-10212539 on Build GRCh38
rs866855203 Chr.X: 10212488-10212488 on Build GRCh38
rs761597049 Chr.X: 10212665-10212665 on Build GRCh38
rs748795719 Chr.X: 10212686-10212686 on Build GRCh38
rs139272007 Chr.X: 10212508-10212508 on Build GRCh38
rs111332758 Chr.X: 10212625-10212625 on Build GRCh38
rs754361706 Chr.X: 10212557-10212557 on Build GRCh38
rs773686595 Chr.X: 10212605-10212605 on Build GRCh38
rs778022533 Chr.X: 10212674-10212674 on Build GRCh38
rs398123627 Chr.X: 10212580-10212580 on Build GRCh38
rs138602675 Chr.X: 10212535-10212535 on Build GRCh38
rs756536616 Chr.X: 10212577-10212577 on Build GRCh38
rs962486915 Chr.X: 10212561-10212561 on Build GRCh38
rs757820245 Chr.X: 10212628-10212628 on Build GRCh38
rs201538709 Chr.X: 10212685-10212685 on Build GRCh38
rs749694905 Chr.X: 10212595-10212595 on Build GRCh38
rs751819949 Chr.X: 10212531-10212531 on Build GRCh38
rs760433125 Chr.X: 10212506-10212506 on Build GRCh38
rs966305132 Chr.X: 10212513-10212513 on Build GRCh38
rs147124557 Chr.X: 10212530-10212530 on Build GRCh38
rs759654800 Chr.X: 10212672-10212672 on Build GRCh38
rs999612580 Chr.X: 10212666-10212666 on Build GRCh38
rs112542881 Chr.X: 10212556-10212556 on Build GRCh38
rs754395139 Chr.X: 10212682-10212682 on Build GRCh38
rs183625504 Chr.X: 10212658-10212658 on Build GRCh38
rs776287214 Chr.X: 10212475-10212475 on Build GRCh38
rs767657932 Chr.X: 10212675-10212675 on Build GRCh38
rs200861909 Chr.X: 10212512-10212512 on Build GRCh38
rs369910318 Chr.X: 10212681-10212681 on Build GRCh38
rs776635142 Chr.X: 10212664-10212664 on Build GRCh38

View Full Product Details