Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41337305-41337534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX3X
Entrez Gene ID
1654
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
UniGene
Hs.728563
Gene Chromosome Location
Chr.X: 41333308-41364472 on Build GRCh38
Transcripts
NM_001193416.2 NM_001193417.2 NM_001356.4 NR_126093.1 NR_126094.1 XM_011543892.2 XM_017029313.1 XM_017029314.1 XM_017030007.1

Gene Symbol
LOC107985678
Entrez Gene ID
107985678
Gene Name
uncharacterized LOC107985678
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.X: 41334304-41337401 on Build GRCh38
Transcripts
NM_001193416.2 NM_001193417.2 NM_001356.4 NR_126093.1 NR_126094.1 XM_011543892.2 XM_017029313.1 XM_017029314.1 XM_017030007.1

SNP Detail

SNP ID Chromosome Location
rs770884779 Chr.X: 41337391-41337391 on Build GRCh38
rs773800236 Chr.X: 41337410-41337410 on Build GRCh38
rs773071639 Chr.X: 41337375-41337375 on Build GRCh38
rs747971521 Chr.X: 41337365-41337365 on Build GRCh38
rs752354649 Chr.X: 41337463-41337463 on Build GRCh38
rs373453343 Chr.X: 41337494-41337494 on Build GRCh38
rs775885529 Chr.X: 41337401-41337401 on Build GRCh38
rs886041769 Chr.X: 41337442-41337443 on Build GRCh38
rs999109544 Chr.X: 41337399-41337399 on Build GRCh38
rs769536255 Chr.X: 41337372-41337372 on Build GRCh38
rs759092210 Chr.X: 41337431-41337431 on Build GRCh38
rs762809683 Chr.X: 41337389-41337389 on Build GRCh38
rs752535289 Chr.X: 41337428-41337429 on Build GRCh38
rs369030204 Chr.X: 41337493-41337493 on Build GRCh38
rs767164397 Chr.X: 41337446-41337446 on Build GRCh38

View Full Product Details