Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172569036-172569305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
UniGene
Hs.204559
Gene Chromosome Location
Chr.1: 172532349-172611833 on Build GRCh38
Transcripts
NM_001282750.1 NM_001282751.1 NM_014283.4 NM_016227.3 XM_006711374.2 XM_006711375.2 XM_006711376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4888722 c.1244C>A p.P415H Chr.1: 172569053-172569053 on Build GRCh38
6212610 c.1297T>A p.W433R Chr.1: 172569106-172569106 on Build GRCh38
6389716 c.844G>A p.E282K Chr.1: 172569130-172569130 on Build GRCh38
3478399 c.1262C>T p.P421L Chr.1: 172569071-172569071 on Build GRCh38
6527892 c.1330A>C p.K444Q Chr.1: 172569139-172569139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765587349 Chr.1: 172569106-172569106 on Build GRCh38
rs1005968244 Chr.1: 172569206-172569206 on Build GRCh38
rs201062317 Chr.1: 172569057-172569057 on Build GRCh38
rs760843676 Chr.1: 172569172-172569172 on Build GRCh38
rs1038427758 Chr.1: 172569263-172569263 on Build GRCh38
rs751438235 Chr.1: 172569192-172569192 on Build GRCh38
rs994290682 Chr.1: 172569278-172569278 on Build GRCh38
rs138977733 Chr.1: 172569111-172569111 on Build GRCh38
rs192029835 Chr.1: 172569271-172569271 on Build GRCh38
rs758667578 Chr.1: 172569160-172569163 on Build GRCh38
rs780853644 Chr.1: 172569062-172569062 on Build GRCh38
rs750710141 Chr.1: 172569141-172569144 on Build GRCh38
rs895124081 Chr.1: 172569203-172569203 on Build GRCh38
rs939265575 Chr.1: 172569161-172569161 on Build GRCh38
rs773305094 Chr.1: 172569124-172569124 on Build GRCh38
rs779965911 Chr.1: 172569080-172569080 on Build GRCh38
rs144718164 Chr.1: 172569117-172569117 on Build GRCh38
rs751583676 Chr.1: 172569168-172569169 on Build GRCh38
rs766535017 Chr.1: 172569169-172569169 on Build GRCh38
rs370898344 Chr.1: 172569118-172569118 on Build GRCh38
rs531909897 Chr.1: 172569071-172569071 on Build GRCh38
rs766623859 Chr.1: 172569178-172569178 on Build GRCh38
rs377016757 Chr.1: 172569093-172569093 on Build GRCh38
rs776955710 Chr.1: 172569181-172569181 on Build GRCh38
rs755516458 Chr.1: 172569178-172569179 on Build GRCh38
rs1052528963 Chr.1: 172569190-172569190 on Build GRCh38
rs982165159 Chr.1: 172569087-172569087 on Build GRCh38
rs373614252 Chr.1: 172569074-172569074 on Build GRCh38

View Full Product Details