Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106201901-106202154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG5
Entrez Gene ID
9474
Gene Name
autophagy related 5
Gene Aliases
APG5, APG5-LIKE, APG5L, ASP, hAPG5
UniGene
Hs.486063
Gene Chromosome Location
Chr.6: 106184476-106325820 on Build GRCh38
Transcripts
NM_001286106.1 NM_001286107.1 NM_001286108.1 NM_001286111.1 NM_004849.3 NR_104402.1 NR_104403.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6452897 c.578C>T p.T193M Chr.6: 106202085-106202085 on Build GRCh38
5987645 c.682G>C p.D228H Chr.6: 106201981-106201981 on Build GRCh38
3766879 c.691+9A>G p.? Chr.6: 106201963-106201963 on Build GRCh38
1487091 c.594C>T p.F198F Chr.6: 106202069-106202069 on Build GRCh38
1311433 c.674C>T p.S225F Chr.6: 106201989-106201989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267600752 Chr.6: 106202053-106202053 on Build GRCh38
rs761593347 Chr.6: 106202101-106202101 on Build GRCh38
rs765857129 Chr.6: 106201940-106201940 on Build GRCh38
rs777898330 Chr.6: 106202043-106202043 on Build GRCh38
rs746396244 Chr.6: 106201947-106201947 on Build GRCh38
rs774715569 Chr.6: 106202096-106202096 on Build GRCh38
rs779280595 Chr.6: 106202052-106202052 on Build GRCh38
rs376589405 Chr.6: 106202048-106202048 on Build GRCh38
rs776478438 Chr.6: 106201991-106201991 on Build GRCh38
rs772578622 Chr.6: 106202075-106202075 on Build GRCh38
rs779600061 Chr.6: 106202077-106202077 on Build GRCh38
rs776190821 Chr.6: 106202097-106202097 on Build GRCh38
rs773158138 Chr.6: 106202117-106202117 on Build GRCh38
rs866038212 Chr.6: 106201996-106201996 on Build GRCh38
rs761736531 Chr.6: 106201992-106201992 on Build GRCh38
rs769649780 Chr.6: 106202115-106202115 on Build GRCh38
rs968989191 Chr.6: 106201937-106201937 on Build GRCh38
rs760314964 Chr.6: 106201983-106201983 on Build GRCh38
rs763708645 Chr.6: 106201984-106201984 on Build GRCh38
rs960601826 Chr.6: 106201965-106201965 on Build GRCh38
rs768319835 Chr.6: 106202089-106202089 on Build GRCh38
rs746306169 Chr.6: 106202055-106202055 on Build GRCh38
rs533087711 Chr.6: 106202027-106202027 on Build GRCh38
rs749760627 Chr.6: 106201953-106201954 on Build GRCh38
rs771529044 Chr.6: 106201950-106201950 on Build GRCh38
rs750350218 Chr.6: 106201993-106201993 on Build GRCh38
rs757567992 Chr.6: 106202050-106202050 on Build GRCh38
rs972289763 Chr.6: 106202000-106202000 on Build GRCh38
rs752779737 Chr.6: 106202033-106202033 on Build GRCh38
rs746564375 Chr.6: 106202085-106202085 on Build GRCh38
rs771397849 Chr.6: 106201943-106201943 on Build GRCh38
rs187511601 Chr.6: 106201946-106201946 on Build GRCh38
rs930548832 Chr.6: 106202037-106202037 on Build GRCh38
rs762972695 Chr.6: 106202118-106202118 on Build GRCh38
rs762992833 Chr.6: 106202021-106202021 on Build GRCh38
rs151260089 Chr.6: 106201954-106201954 on Build GRCh38
rs756357015 Chr.6: 106202038-106202038 on Build GRCh38
rs1027810207 Chr.6: 106202036-106202036 on Build GRCh38
rs373882027 Chr.6: 106202026-106202026 on Build GRCh38
rs374402837 Chr.6: 106202119-106202119 on Build GRCh38
rs768920976 Chr.6: 106201965-106201966 on Build GRCh38

View Full Product Details