Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58115220-58115493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP20
Entrez Gene ID
29105
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, EVORF, GTL3, fSAP23
UniGene
Hs.532755
Gene Chromosome Location
Chr.16: 58113588-58129425 on Build GRCh38
Transcripts
NM_013242.2 NR_136632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614583 c.433G>A p.G145S Chr.16: 58115301-58115301 on Build GRCh38
4490202 c.359C>T p.T120I Chr.16: 58115375-58115375 on Build GRCh38
471871 c.344A>G p.K115R Chr.16: 58115390-58115390 on Build GRCh38
1198203 c.298C>T p.R100C Chr.16: 58115436-58115436 on Build GRCh38
4879348 c.435C>G p.G145G Chr.16: 58115299-58115299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762892615 Chr.16: 58115272-58115272 on Build GRCh38
rs752101250 Chr.16: 58115423-58115423 on Build GRCh38
rs764092647 Chr.16: 58115394-58115394 on Build GRCh38
rs144778705 Chr.16: 58115353-58115353 on Build GRCh38
rs191235633 Chr.16: 58115245-58115245 on Build GRCh38
rs773775681 Chr.16: 58115261-58115261 on Build GRCh38
rs200960487 Chr.16: 58115352-58115352 on Build GRCh38
rs554565830 Chr.16: 58115368-58115368 on Build GRCh38
rs914251254 Chr.16: 58115424-58115424 on Build GRCh38
rs774639242 Chr.16: 58115251-58115251 on Build GRCh38
rs767845554 Chr.16: 58115397-58115397 on Build GRCh38
rs767826166 Chr.16: 58115301-58115301 on Build GRCh38
rs757595047 Chr.16: 58115294-58115294 on Build GRCh38
rs746684277 Chr.16: 58115254-58115254 on Build GRCh38
rs775019974 Chr.16: 58115374-58115374 on Build GRCh38
rs755182323 Chr.16: 58115337-58115337 on Build GRCh38
rs779278514 Chr.16: 58115362-58115362 on Build GRCh38
rs748479016 Chr.16: 58115364-58115364 on Build GRCh38
rs769205282 Chr.16: 58115372-58115372 on Build GRCh38
rs750612612 Chr.16: 58115419-58115419 on Build GRCh38
rs750572831 Chr.16: 58115302-58115302 on Build GRCh38
rs755515452 Chr.16: 58115455-58115455 on Build GRCh38
rs538753443 Chr.16: 58115246-58115246 on Build GRCh38
rs761689275 Chr.16: 58115396-58115396 on Build GRCh38
rs1016266568 Chr.16: 58115238-58115238 on Build GRCh38
rs571323556 Chr.16: 58115243-58115243 on Build GRCh38
rs200407739 Chr.16: 58115305-58115305 on Build GRCh38
rs78940528 Chr.16: 58115460-58115460 on Build GRCh38
rs781373557 Chr.16: 58115467-58115468 on Build GRCh38
rs181230226 Chr.16: 58115287-58115287 on Build GRCh38
rs765289914 Chr.16: 58115263-58115263 on Build GRCh38
rs576289782 Chr.16: 58115380-58115380 on Build GRCh38
rs1031146995 Chr.16: 58115435-58115435 on Build GRCh38
rs770616964 Chr.16: 58115367-58115367 on Build GRCh38
rs780112642 Chr.16: 58115310-58115310 on Build GRCh38
rs745461080 Chr.16: 58115371-58115371 on Build GRCh38
rs764778355 Chr.16: 58115355-58115355 on Build GRCh38
rs779368456 Chr.16: 58115462-58115462 on Build GRCh38
rs753911484 Chr.16: 58115436-58115436 on Build GRCh38
rs774338432 Chr.16: 58115395-58115395 on Build GRCh38
rs371674432 Chr.16: 58115432-58115432 on Build GRCh38
rs141032801 Chr.16: 58115269-58115269 on Build GRCh38
rs930058372 Chr.16: 58115253-58115253 on Build GRCh38
rs1025217182 Chr.16: 58115257-58115257 on Build GRCh38
rs185660863 Chr.16: 58115422-58115422 on Build GRCh38
rs770529103 Chr.16: 58115255-58115255 on Build GRCh38
rs764006782 Chr.16: 58115286-58115286 on Build GRCh38

View Full Product Details