Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44666966-44667239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIOC
Entrez Gene ID
284071
Gene Name
meiosis specific with coiled-coil domain
Gene Aliases
C17orf104
UniGene
Hs.349758
Gene Chromosome Location
Chr.17: 44656394-44677086 on Build GRCh38
Transcripts
NM_001145080.2 XM_005257236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6857817 c.1162G>T p.D388Y Chr.17: 44667073-44667073 on Build GRCh38
7358130 c.1229C>G p.A410G Chr.17: 44667140-44667140 on Build GRCh38
6992667 c.1191T>A p.T397T Chr.17: 44667102-44667102 on Build GRCh38
3191330 c.1293C>T p.P431P Chr.17: 44667204-44667204 on Build GRCh38
374247 c.1105A>T p.T369S Chr.17: 44667016-44667016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776619348 Chr.17: 44667045-44667045 on Build GRCh38
rs779431275 Chr.17: 44667029-44667029 on Build GRCh38
rs367775822 Chr.17: 44667204-44667204 on Build GRCh38
rs1037688635 Chr.17: 44667121-44667121 on Build GRCh38
rs752597911 Chr.17: 44667051-44667051 on Build GRCh38
rs765083876 Chr.17: 44667049-44667049 on Build GRCh38
rs772628987 Chr.17: 44667160-44667160 on Build GRCh38
rs771266675 Chr.17: 44667042-44667042 on Build GRCh38
rs762890394 Chr.17: 44667054-44667054 on Build GRCh38
rs946466055 Chr.17: 44667143-44667143 on Build GRCh38
rs376561059 Chr.17: 44667104-44667104 on Build GRCh38
rs368090785 Chr.17: 44667041-44667041 on Build GRCh38
rs910287376 Chr.17: 44667183-44667183 on Build GRCh38
rs751077830 Chr.17: 44667209-44667209 on Build GRCh38
rs372239891 Chr.17: 44667055-44667055 on Build GRCh38
rs777022275 Chr.17: 44667194-44667194 on Build GRCh38
rs764075829 Chr.17: 44667207-44667207 on Build GRCh38
rs777288104 Chr.17: 44666995-44666995 on Build GRCh38
rs757875577 Chr.17: 44666993-44666993 on Build GRCh38
rs189011609 Chr.17: 44667112-44667112 on Build GRCh38
rs759372564 Chr.17: 44667047-44667047 on Build GRCh38
rs780789280 Chr.17: 44667057-44667059 on Build GRCh38
rs751184425 Chr.17: 44666996-44666996 on Build GRCh38
rs749636749 Chr.17: 44667013-44667013 on Build GRCh38
rs747065517 Chr.17: 44667176-44667176 on Build GRCh38
rs777775029 Chr.17: 44667167-44667167 on Build GRCh38
rs759782330 Chr.17: 44667195-44667195 on Build GRCh38
rs374546581 Chr.17: 44667006-44667006 on Build GRCh38
rs752289652 Chr.17: 44666991-44666991 on Build GRCh38
rs1051813290 Chr.17: 44667060-44667060 on Build GRCh38
rs889194976 Chr.17: 44667061-44667061 on Build GRCh38
rs769724391 Chr.17: 44667196-44667196 on Build GRCh38
rs771321655 Chr.17: 44667186-44667186 on Build GRCh38
rs748612319 Chr.17: 44667147-44667147 on Build GRCh38
rs769194089 Chr.17: 44667021-44667021 on Build GRCh38
rs750040547 Chr.17: 44667113-44667113 on Build GRCh38
rs780460643 Chr.17: 44667009-44667009 on Build GRCh38
rs60072626 Chr.17: 44667105-44667105 on Build GRCh38
rs144880520 Chr.17: 44667140-44667140 on Build GRCh38
rs779282328 Chr.17: 44667145-44667145 on Build GRCh38
rs748644023 Chr.17: 44667030-44667030 on Build GRCh38

View Full Product Details