Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5922351-5922610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHGB
Entrez Gene ID
1114
Gene Name
chromogranin B
Gene Aliases
SCG1
UniGene
Hs.516874
Gene Chromosome Location
Chr.20: 5911328-5925361 on Build GRCh38
Transcripts
NM_001819.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
402196 c.412C>A p.Q138K Chr.20: 5922556-5922556 on Build GRCh38
4544194 c.351G>A p.K117K Chr.20: 5922495-5922495 on Build GRCh38
149134 c.277T>A p.S93T Chr.20: 5922421-5922421 on Build GRCh38
4541689 c.304G>A p.G102R Chr.20: 5922448-5922448 on Build GRCh38
3548453 c.412C>T p.Q138* Chr.20: 5922556-5922556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200903465 Chr.20: 5922536-5922536 on Build GRCh38
rs199738388 Chr.20: 5922381-5922381 on Build GRCh38
rs918150134 Chr.20: 5922537-5922537 on Build GRCh38
rs376991086 Chr.20: 5922422-5922422 on Build GRCh38
rs144110872 Chr.20: 5922490-5922490 on Build GRCh38
rs762563946 Chr.20: 5922555-5922555 on Build GRCh38
rs745742066 Chr.20: 5922507-5922507 on Build GRCh38
rs879912575 Chr.20: 5922500-5922500 on Build GRCh38
rs951929950 Chr.20: 5922418-5922418 on Build GRCh38
rs774728448 Chr.20: 5922404-5922404 on Build GRCh38
rs751378560 Chr.20: 5922570-5922570 on Build GRCh38
rs768860358 Chr.20: 5922514-5922514 on Build GRCh38
rs748225368 Chr.20: 5922589-5922589 on Build GRCh38
rs764672301 Chr.20: 5922441-5922441 on Build GRCh38
rs752205651 Chr.20: 5922553-5922553 on Build GRCh38
rs6085324 Chr.20: 5922421-5922421 on Build GRCh38
rs374038627 Chr.20: 5922432-5922432 on Build GRCh38
rs761132826 Chr.20: 5922430-5922430 on Build GRCh38
rs773068018 Chr.20: 5922499-5922499 on Build GRCh38
rs752353688 Chr.20: 5922449-5922449 on Build GRCh38
rs142077939 Chr.20: 5922542-5922542 on Build GRCh38
rs200658508 Chr.20: 5922520-5922520 on Build GRCh38
rs749349645 Chr.20: 5922387-5922387 on Build GRCh38
rs779181544 Chr.20: 5922586-5922586 on Build GRCh38
rs766238400 Chr.20: 5922461-5922461 on Build GRCh38
rs11558194 Chr.20: 5922531-5922531 on Build GRCh38
rs236150 Chr.20: 5922495-5922495 on Build GRCh38
rs776182778 Chr.20: 5922569-5922569 on Build GRCh38
rs973757064 Chr.20: 5922511-5922511 on Build GRCh38
rs891947037 Chr.20: 5922541-5922541 on Build GRCh38
rs368450620 Chr.20: 5922576-5922576 on Build GRCh38
rs78101688 Chr.20: 5922546-5922546 on Build GRCh38
rs777454124 Chr.20: 5922485-5922485 on Build GRCh38
rs184354829 Chr.20: 5922574-5922574 on Build GRCh38
rs367915171 Chr.20: 5922491-5922491 on Build GRCh38
rs138069166 Chr.20: 5922423-5922423 on Build GRCh38
rs547704720 Chr.20: 5922433-5922433 on Build GRCh38
rs748338597 Chr.20: 5922525-5922525 on Build GRCh38
rs957159617 Chr.20: 5922393-5922393 on Build GRCh38
rs758031352 Chr.20: 5922466-5922466 on Build GRCh38
rs756065519 Chr.20: 5922579-5922579 on Build GRCh38
rs750417629 Chr.20: 5922578-5922578 on Build GRCh38
rs867120304 Chr.20: 5922563-5922563 on Build GRCh38
rs6133278 Chr.20: 5922577-5922577 on Build GRCh38
rs371451030 Chr.20: 5922427-5922427 on Build GRCh38
rs917291982 Chr.20: 5922458-5922458 on Build GRCh38
rs759973072 Chr.20: 5922547-5922547 on Build GRCh38
rs375812326 Chr.20: 5922545-5922545 on Build GRCh38
rs950031427 Chr.20: 5922476-5922476 on Build GRCh38
rs764761970 Chr.20: 5922548-5922548 on Build GRCh38
rs567433214 Chr.20: 5922526-5922526 on Build GRCh38
rs766836438 Chr.20: 5922431-5922431 on Build GRCh38
rs148482419 Chr.20: 5922399-5922399 on Build GRCh38

View Full Product Details