Gene Transcripts

Gene Symbol
Mapre3
Entrez Gene ID
100732
Gene Name
microtubule-associated protein, RP/EB family, member 3
Gene Aliases
AI790651, EB2, EB3
Chromosome Location
Chr. 5: 30814614 - 30866106 on Build GRCm38
UniGene ID
Mm.332295
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133350.1 NP_579928.1 4 580
GenBank mRNA AF468005.1 - 4 580
BC048833.1 - 3 1142
BC057918.1 - 4 580 5134461
U51204.1 - 2 262

View Full Product Details