Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029770.2 | NP_084046.2 | 14 | 2644 | |
GenBank mRNA | AJ487853.1 | - | 14 | 2547 | |
AK018177.1 | - | 14 | 2644 | ||
AK147603.1 | - | 13 | 2506 | ||
AK154271.1 | - | 14 | 2644 | ||
AK154287.1 | - | 14 | 2648 | ||
AK158284.1 | - | 14 | 2644 | ||
BC048162.1 | - | 13 | 2418 | ||
BC057560.1 | - | 13 | 2418 | 6417563 |