Gene Transcripts

Gene Symbol
Bcl11a
Entrez Gene ID
14025
Gene Name
B cell CLL/lymphoma 11A (zinc finger protein)
Gene Aliases
2810047E18Rik, BCL-11A, Ctip1, D930021L15Rik, Evi9, Evi9a, Evi9b, Evi9c, mKIAA1809
Chromosome Location
Chr. 11: 24076564 - 24173558 on Build GRCm38
UniGene ID
Mm.53687
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159289.1 NP_001152761.1 5 1036
NM_001159290.1 NP_001152762.1 5 691
NM_016707.3 NP_057916.1 5 2636
GenBank mRNA AF051525.1 - 5 2574
AF169036.1 - 3 1777
AF169037.1 - 5 975
AF186018.1 - 5 2516
AK004395.1 - 5 912
AK043677.1 - 5 835
AK045556.1 - 5 692
BC010585.1 - 2 1650 2631265
BC051418.1 - 5 824

Gene Symbol
4930538E20Rik
Entrez Gene ID
78200
Gene Name
RIKEN cDNA 4930538E20 gene
Gene Aliases
Chromosome Location
Chr. 11: 24159903 - 24172824 on Build GRCm38
UniGene ID
Mm.455965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK019630.1 - 1 53

View Full Product Details