Gene Transcripts

Gene Symbol
Ppm1g
Entrez Gene ID
14208
Gene Name
protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform
Gene Aliases
AI385652, Fin13
Chromosome Location
Chr. 5: 31202668 - 31220545 on Build GRCm38
UniGene ID
Mm.14501
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008014.3 NP_032040.1 9 1524
GenBank mRNA AK187638.1 - 3 335
BC009004.1 - 9 1524 3499081
U42383.1 - 9 1410

View Full Product Details