Gene Transcripts

Gene Symbol
Asb13
Entrez Gene ID
142688
Gene Name
ankyrin repeat and SOCS box-containing 13
Gene Aliases
2210015B19Rik, 6430573K02Rik, Asb-13, C85285
Chromosome Location
Chr. 13: 3634012 - 3651779 on Build GRCm38
UniGene ID
Mm.253061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178283.4 NP_840068.1 3 434
GenBank mRNA AF403041.1 - 3 434
AK078289.1 - 3 390
AK133892.1 - 3 360
BC018240.1 - 3 388 4239996

View Full Product Details