Gene Transcripts
- Gene Symbol
- Arhgap4
- Entrez Gene ID
- 171207
- Gene Name
- Rho GTPase activating protein 4
- Gene Aliases
- A130009C12Rik, A530015A20Rik, Rgc1, c1, mKIAA0131
- Chromosome Location
- Chr. X: 73894352 - 73911346 on Build GRCm38
- UniGene ID
- Mm.482337
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001162423.1 |
NP_001155895.1
|
19, 20 |
2494 |
|
NM_001162424.1 |
NP_001155896.1
|
18, 19 |
2464 |
|
NM_138630.2 |
NP_619536.2
|
19, 20 |
2527 |
|
GenBank mRNA |
AF443826.1 |
- |
19, 20 |
2458 |
|
AK040687.1 |
- |
12, 13 |
1333 |
|
AK129060.1 |
- |
|
3036 |
|
AK157294.1 |
- |
19, 20 |
2526 |
|
AK169865.1 |
- |
16, 17 |
1962 |
|
AY183124.1 |
- |
19, 20 |
2500 |
|
AY183125.1 |
- |
18, 19 |
2437 |
|
AY183126.1 |
- |
18, 19 |
2594 |
|
AY183127.1 |
- |
19, 20 |
2500 |
|
AY183128.1 |
- |
15, 16 |
2189 |
|
AY183129.1 |
- |
19, 20 |
2500 |
|
AY183130.1 |
- |
15, 16 |
1872 |
|
AY183131.1 |
- |
14, 15 |
1966 |
|
AY183132.1 |
- |
13, 14 |
2192 |
|
AY183133.1 |
- |
17, 18 |
2533 |
|
BC109001.1 |
- |
19, 20 |
2425 |
40048081 |
BC109002.1 |
- |
17, 18 |
2484 |
|
View Full Product Details