Gene Transcripts

Gene Symbol
Olfr52
Entrez Gene ID
18352
Gene Name
olfactory receptor 52
Gene Aliases
IE6, MOR185-6, ORL446
Chromosome Location
Chr. 2: 86181150 - 86182109 on Build GRCm38
UniGene ID
Mm.222842
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146583.1 NP_666794.1 1 610
GenBank mRNA AF102528.1 - 1 421

View Full Product Details