Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252453.1 | NP_001239382.1 | 9 | 1846 | |
NM_001252455.1 | NP_001239384.1 | 9 | 1846 | ||
NM_001252456.1 | NP_001239385.1 | 9 | 1823 | ||
NM_011218.2 | NP_035348.2 | 9 | 1846 | ||
GenBank mRNA | BC052462.1 | - | 9 | 1772 | |
BC054077.1 | - | 9 | 1740 | ||
BC083188.1 | - | 9 | 1805 | 5709264 | |
D28530.1 | - | 9 | 1735 | ||
D28531.1 | - | 9 | 1735 | ||
X82288.1 | - | 8 | 1655 |