Gene Transcripts

Gene Symbol
Hoxc10
Entrez Gene ID
209448
Gene Name
homeobox C10
Gene Aliases
AI506621, Hox-3.6
Chromosome Location
Chr. 15: 102966796 - 102971898 on Build GRCm38
UniGene ID
Mm.31665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010462.5 NP_034592.2 1 401
GenBank mRNA AK033186.1 - 1 333
AK138664.1 - 1 387
BC053405.1 - 1 413 30059581

View Full Product Details