Gene Transcripts

Gene Symbol
Chrm5
Entrez Gene ID
213788
Gene Name
cholinergic receptor, muscarinic 5
Gene Aliases
AChR-M5, M5R, m5
Chromosome Location
Chr. 2: 112478733 - 112537563 on Build GRCm38
UniGene ID
Mm.297624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205783.2 NP_991352.2 1 918
GenBank mRNA BC120615.1 - 1 918

View Full Product Details