Gene Transcripts

Gene Symbol
Mis18bp1
Entrez Gene ID
217653
Gene Name
MIS18 binding protein 1
Gene Aliases
6720403H11, C79407, Knl2, mKIAA1903
Chromosome Location
Chr. 12: 65132734 - 65172580 on Build GRCm38
UniGene ID
Mm.21144
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172578.2 NP_766166.2 8 2429
GenBank mRNA AK032698.1 - 6 1461
AK035117.1 - 8 2428
AK220223.1 - 7 1695
BC021832.1 - 3 942
BC052175.1 - 8 2429 6394068

View Full Product Details