Gene Transcripts

Gene Symbol
Tst
Entrez Gene ID
22117
Gene Name
thiosulfate sulfurtransferase, mitochondrial
Gene Aliases
Rhodanese
Chromosome Location
Chr. 15: 78399556 - 78405859 on Build GRCm38
UniGene ID
Mm.15312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009437.4 NP_033463.1 1 230
GenBank mRNA AK003953.1 - 1 237
BC005644.1 - 1 254 3257034
CT010210.1 - 1 204
U35741.1 - 1 218

View Full Product Details