Gene Transcripts

Gene Symbol
Bag6
Entrez Gene ID
224727
Gene Name
BCL2-associated athanogene 6
Gene Aliases
2410045D21Rik, AA408914, BAG-6, Bat3, D17H6S52E, G3, Scythe
Chromosome Location
Chr. 17: 35134600 - 35147322 on Build GRCm38
UniGene ID
Mm.203962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252468.1 NP_001239397.1 23, 24 3627
NM_057171.2 NP_476512.1 24, 25 3681
GenBank mRNA AK010670.1 - 2, 3 255
AK148796.1 - 23, 24 3547
BC026647.1 - 16, 17 2422
BC030296.1 - 5, 6 550
BY595553.1 - 3, 4 286

View Full Product Details