Gene Transcripts

Gene Symbol
Mapkap1
Entrez Gene ID
227743
Gene Name
mitogen-activated protein kinase associated protein 1
Gene Aliases
AI591529, D230039K05Rik, Sin1
Chromosome Location
Chr. 2: 34406771 - 34624958 on Build GRCm38
UniGene ID
Mm.270866
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290625.1 NP_001277554.1 2, 3 416
NM_001290626.1 NP_001277555.1 1, 2 414
NM_177345.4 NP_796319.1 2, 3 742
GenBank mRNA AK042154.1 - 1, 2 404
AK052045.1 - 2, 3 410
AK132263.1 - 2, 3 406
AK148428.1 - 2, 3 731
BC043296.1 - 2, 3 641
BC090644.1 - 1, 2 316 30629263
BC096618.1 - 2, 3 715 6392317

View Full Product Details