Gene Transcripts

Gene Symbol
Brat1
Entrez Gene ID
231841
Gene Name
BRCA1-associated ATM activator 1
Gene Aliases
AA881470, Baat1
Chromosome Location
Chr. 5: 140704786 - 140719379 on Build GRCm38
UniGene ID
Mm.271932
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276287.1 NP_001263216.1 13 1844
NM_181066.3 NP_851411.1 13 1746
GenBank mRNA AF226663.1 - 3 293
AK049321.1 - 12 1643
AK084987.1 - 13 1735
AK085091.1 - 13 1735
AK154448.1 - 13 1717
AK170269.1 - 13 1740
BC068301.1 - 13 1810 30061848

View Full Product Details