Gene Transcripts

Gene Symbol
Spry3
Entrez Gene ID
236576
Gene Name
sprouty homolog 3 (Drosophila)
Gene Aliases
Gm1409, Gm391, sprouty3
Chromosome Location
Chr. X|NT_165789: 101352 - 110912 on Build GRCm38
UniGene ID
Mm.297666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030293.2 NP_001025464.1 2 503
GenBank mRNA AK138539.1 - 2 503
BC132135.1 - 1 118 40130412
BC132137.1 - 1 118 40130414

View Full Product Details