Gene Transcripts

Gene Symbol
Mab21l2
Entrez Gene ID
23937
Gene Name
mab-21-like 2 (C. elegans)
Gene Aliases
Chromosome Location
Chr. 3: 86545581 - 86548283 on Build GRCm38
UniGene ID
Mm.389466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011839.3 NP_035969.2 1 618
GenBank mRNA AF126427.1 - 1 959
AF149877.1 - 1 360
AK053645.1 - 1 619
AK144373.1 - 1 786
BC046628.1 - 1 584 6493460
BC058730.1 - 1 724

View Full Product Details