Gene Transcripts

Gene Symbol
Sh3bp5
Entrez Gene ID
24056
Gene Name
SH3-domain binding protein 5 (BTK-associated)
Gene Aliases
AI606498, SH3BP-5, Sab
Chromosome Location
Chr. 14: 31373774 - 31436033 on Build GRCm38
UniGene ID
Mm.383198
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011894.2 NP_036024.2 2 245
GenBank mRNA AB016835.1 - 2 73
AK043375.1 - 290
AK165033.1 - 2 279
BC018237.1 - 2 277 3482670
BC053741.1 - 2 245 6336511

View Full Product Details