Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145700.2 | NP_663746.2 | 3 | 1077 | |
GenBank mRNA | AF306532.1 | - | 1 | 626 | |
AK030643.1 | - | 2 | 643 | ||
AK042430.1 | - | 2 | 748 | ||
AY072796.1 | - | 2 | 1328 | ||
AY072938.1 | - | 3 | 1369 | ||
BC140999.1 | - | 3 | 1077 | 9055611 |