Gene Transcripts

Gene Symbol
Mapk13
Entrez Gene ID
26415
Gene Name
mitogen-activated protein kinase 13
Gene Aliases
SAPK4, Serk4
Chromosome Location
Chr. 17: 28769317 - 28778704 on Build GRCm38
UniGene ID
Mm.27970
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011950.2 NP_036080.2 9, 10 864
GenBank mRNA AK146993.1 - 9, 10 852
BC001992.1 - 9, 10 833 3492903
U81823.1 - 9, 10 821

View Full Product Details