Gene Transcripts

Gene Symbol
Ccdc27
Entrez Gene ID
381580
Gene Name
coiled-coil domain containing 27
Gene Aliases
Gm1035
Chromosome Location
Chr. 4: 154026644 - 154042982 on Build GRCm38
UniGene ID
Mm.51001
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033455.2 NP_001028627.1 4 521
GenBank mRNA AK133459.1 - 4 522
BC150930.1 - 4 504 9087841
BC151167.1 - 4 504 9088056

View Full Product Details