Gene Transcripts

Gene Symbol
C7
Entrez Gene ID
109828
Gene Name
complement component 7
Gene Aliases
A530026G17, Gm1238
Chromosome Location
Chr. 15: 4988762 - 5063773 on Build GRCm38
UniGene ID
Mm.208034
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243837.1 NP_001230766.1 14 1977

View Full Product Details