Gene Transcripts

Gene Symbol
Ifnk
Entrez Gene ID
387510
Gene Name
interferon kappa
Gene Aliases
Chromosome Location
Chr. 4: 35152055 - 35154019 on Build GRCm38
UniGene ID
Mm.386929
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199157.2 NP_954608.1 1 243
GenBank mRNA BC099376.1 - 1 243

View Full Product Details