Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134257.3 | NP_599018.3 | 12, 13 | 2453 | |
GenBank mRNA | AF215670.1 | - | 2, 3 | 384 | |
AF350047.1 | - | 13, 14 | 2328 | ||
AK004648.1 | - | 6, 7 | 1622 | ||
BC033449.1 | - | 3, 4 | 1469 | 3599345 |