Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252463.1 | NP_001239392.1 | 2 | 197 | |
NM_001252464.1 | NP_001239393.1 | 2 | 172 | ||
NM_019988.5 | NP_064372.2 | 2 | 192 | ||
GenBank mRNA | AF237676.1 | - | 2 | 123 | |
AK002751.1 | - | 2 | 158 | ||
AK038515.1 | - | 1 | 690 | ||
AK040100.1 | - | 2 | 193 | ||
AK048102.1 | - | 2 | 156 | ||
AK077680.1 | - | 2 | 163 | ||
AK083731.1 | - | 2 | 133 | ||
AK088951.1 | - | 2 | 165 | ||
BC015279.1 | - | 2 | 151 | 4222934 | |
CF750475.1 | - | 2 | 161 |