Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020580.3 | NP_065605.2 | 8 | 998 | |
GenBank mRNA | AJ238741.1 | - | 8 | 932 | |
AJ238742.1 | - | 8 | 998 | ||
AJ238743.1 | - | 8 | 952 | ||
AK010360.1 | - | 8 | 945 | ||
AK049418.1 | - | 8 | 970 | ||
AK159866.1 | - | 8 | 999 | ||
AK184518.1 | - | 2 | 221 | ||
BC006959.1 | - | 8 | 959 | 3707929 | |
BC031747.1 | - | 8 | 956 | 3673038 |