Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146211.1 | NP_001139683.1 | 6 | 491 | |
NM_001146212.1 | NP_001139684.1 | 6 | 494 | ||
NM_183086.2 | NP_898909.2 | 6 | 584 | ||
GenBank mRNA | AK004151.1 | - | 6 | 488 | |
AK136970.1 | - | 2 | 616 | ||
BC038917.1 | - | 6 | 480 | ||
BC048912.1 | - | 6 | 587 | 4238014 | |
BC096043.1 | - | 5 | 449 | ||
BC096657.1 | - | 5 | 399 | 5144028 | |
DV046794.1 | - | 6 | 505 |