Gene Transcripts

Gene Symbol
Atg10
Entrez Gene ID
66795
Gene Name
autophagy related 10
Gene Aliases
5330424L23Rik, 5430428K15Rik, AI852123, APG10, Agp10, Apg10l, Apg10p, Atg10l
Chromosome Location
Chr. 13: 90935348 - 91224792 on Build GRCm38
UniGene ID
Mm.235385
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025770.3 NP_080046.3 4 382
GenBank mRNA AB079383.1 - 3 338
AB091691.1 - 3 359
AK009076.1 - 4 366
AK017355.1 - 4 364
AK077333.1 - 4 355
BC023455.1 - 3 358 5249268

View Full Product Details